Merck
  • Structure-function relationships of the soluble form of the antiaging protein Klotho have therapeutic implications for managing kidney disease.
Structure-function relationships of the soluble form of the antiaging protein Klotho have therapeutic implications for managing kidney disease.
The Journal of biological chemistry (2020-02-02)
Xiaotian Zhong, Srinath Jagarlapudi, Yan Weng, Mellisa Ly, Jason C Rouse, Kim McClure, Tetsuya Ishino, Yan Zhang, Eric Sousa, Justin Cohen, Boriana Tzvetkova, Kaffa Cote, John J Scarcelli, Keith Johnson, Joe Palandra, James R Apgar, Suma Yaddanapudi, Romer A Gonzalez-Villalobos, Alan C Opsahl, Khetemenee Lam, Qing Yao, Weili Duan, Annette Sievers, Jing Zhou, Darren Ferguson, Aaron D'Antona, Richard Zollner, Hongli L Zhu, Ron Kriz, Laura Lin, Valerie Clerin
ABSTRACT
The fortuitously discovered antiaging membrane protein αKlotho (Klotho) is highly expressed in the kidney, and deletion of the Klotho gene in mice causes a phenotype strikingly similar to that of chronic kidney disease (CKD). Klotho functions as a co-receptor for fibroblast growth factor 23 (FGF23) signaling, whereas its shed extracellular domain, soluble Klotho (sKlotho), carrying glycosidase activity, is a humoral factor that regulates renal health. Low sKlotho in CKD is associated with disease progression, and sKlotho supplementation has emerged as a potential therapeutic strategy for managing CKD. Here, we explored the structure-function relationship and post-translational modifications of sKlotho variants to guide the future design of sKlotho-based therapeutics. Chinese hamster ovary (CHO)- and human embryonic kidney (HEK)-derived WT sKlotho proteins had varied activities in FGF23 co-receptor and β-glucuronidase assays in vitro and distinct properties in vivo Sialidase treatment of heavily sialylated CHO-sKlotho increased its co-receptor activity 3-fold, yet it remained less active than hyposialylated HEK-sKlotho. MS and glycopeptide-mapping analyses revealed that HEK-sKlotho is uniquely modified with an unusual N-glycan structure consisting of N,N'-di-N-acetyllactose diamine at multiple N-linked sites, one of which at Asn-126 was adjacent to a putative GalNAc transfer motif. Site-directed mutagenesis and structural modeling analyses directly implicated N-glycans in Klotho's protein folding and function. Moreover, the introduction of two catalytic glutamate residues conserved across glycosidases into sKlotho enhanced its glucuronidase activity but decreased its FGF23 co-receptor activity, suggesting that these two functions might be structurally divergent. These findings open up opportunities for rational engineering of pharmacologically enhanced sKlotho therapeutics for managing kidney disease.
Sigma-Aldrich
β-Glucuronidase from bovine liver, Type B-1, ≥1,000,000 units/g solid
Sigma-Aldrich
4-Methylumbelliferyl-β-D-glucuronide hydrate, ≥98% (HPLC), BioReagent, for identification of transformed plants