Merck
  • Temperature and storage time increase provitamin A carotenoid concentrations and bioaccessibility in post-harvest carrots.
Temperature and storage time increase provitamin A carotenoid concentrations and bioaccessibility in post-harvest carrots.
Food chemistry (2020-09-21)
Faiza Hammaz, Florence Charles, Rachel E Kopec, Charlotte Halimi, Salah Fgaier, Jawad Aarrouf, Laurent Urban, Patrick Borel
ABSTRACT
The aim was to enhance provitamin A carotenoid (proVA CAR) concentrations and bioaccessibility in carrots by manipulating post-harvest factors. To that end, we assessed the effects of Ultraviolet-C light, pulsed light, storage temperature, and storage duration. We also measured CAR bioaccessibility by using an in vitro model. Pulsed light, but not Ultraviolet-C, treatment increased proVA CAR concentrations in the cortex tissue (pā€Æ<ā€Æ0.05). Longer storage times and higher temperatures also increased concentrations (pā€Æ<ā€Æ0.05). The maximal increase induced by pulsed light was obtained after treatment with 20ā€ÆkJ/m2 and 3-days of storage at 20ā€ÆĀ°C. However, the positive effect induced by pulsed light decreased considerably over the next seven days. ProVA CAR in carrots with the highest concentrations also proved to be more bioaccessible (pā€Æ<ā€Æ0.05). Thus, proVA CAR concentrations in stored carrots can be increased significantly through storage times and temperatures. Pulsed light can also significantly increase proVA CAR concentrations, but only temporarily.
Sigma-Aldrich
Echinenone, ≥95.0% (HPLC)