Merck
  • Photo-responsive supramolecular hyaluronic acid hydrogels for accelerated wound healing.
Photo-responsive supramolecular hyaluronic acid hydrogels for accelerated wound healing.
Journal of controlled release : official journal of the Controlled Release Society (2020-04-14)
Weiyi Zhao, Yang Li, Xiang Zhang, Rui Zhang, Yang Hu, Cyrille Boyer, Fu-Jian Xu
ABSTRACT
Supramolecular hydrogels confer control over structural properties in a reversible, dynamic, and biomimetic fashion. The design of supramolecular hydrogels with an improved structural and functional recapitulation of damaged organs is important for clinical applications. For wound healing management, in particular, an effective healing process, through the modulation of epidermal growth factor (EGF) delivery using supramolecular polysaccharide hydrogels, has yet to be developed. In this study, photo-responsive supramolecular polysaccharide hydrogels were formed through host-guest interactions between azobenzene and β-cyclodextrin groups conjugated to hyaluronic acid chains. By exploiting the photoisomerization properties of azobenzene under different wavelengths, a supramolecular hydrogel featuring a dynamic spatial network crosslink density through the application of a light stimulus was obtained. Under ultra violet (UV) light, the loosened hydrogel can rapidly release EGF, thereby enhancing EGF delivery at the wound site. Based on an in vivo assessment of the healing process through a full-thickness skin defect model, the controlled EGF release from a supramolecular hydrogel exhibited superior wound healing efficiency with respect to granulation tissue formation, growth factor levels, and angiogenesis. Therefore, the proposed supramolecular hydrogels are potentially valuable as controlled delivery systems for future clinical wound healing applications.
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
1-Adamantanecarbonyl chloride, 95%