Merck
  • Intestine-selective reduction of Gcg expression reveals the importance of the distal gut for GLP-1 secretion.
Intestine-selective reduction of Gcg expression reveals the importance of the distal gut for GLP-1 secretion.
Molecular metabolism (2020-04-13)
Brandon L Panaro, Bernardo Yusta, Dianne Matthews, Jacqueline A Koehler, Youngmi Song, Darleen A Sandoval, Daniel J Drucker
ABSTRACT
Glucagon-like peptide-1 is a nutrient-sensitive hormone secreted from enteroendocrine L cells within the small and large bowel. Although GLP-1 levels rise rapidly in response to food ingestion, the greatest density of L cells is localized to the distal small bowel and colon. Here, we assessed the importance of the distal gut in the acute L cell response to diverse secretagogues. Circulating levels of glucose and plasma GLP-1 were measured in response to the administration of L cell secretagogues in wild-type mice and in mice with (1) genetic reduction of Gcg expression throughout the small bowel and large bowel (GcgGut-/-) and (2) selective reduction of Gcg expression in the distal gut (GcgDistalGut-/-). The acute GLP-1 response to olive oil or arginine administration was markedly diminished in GcgGut-/- but preserved in GcgDistalGut-/- mice. In contrast, the increase in plasma GLP-1 levels following the administration of the GPR119 agonist AR231453, or the melanocortin-4 receptor (MC4R) agonist LY2112688, was markedly diminished in the GcgDistalGut-/- mice. The GLP-1 response to LPS was also markedly attenuated in the GcgGut-/- mice and remained submaximal in the GcgDistalGut-/- mice. Doses of metformin sufficient to lower glucose and increase GLP-1 levels in the GcgGut+/+ mice retained their glucoregulatory activity, yet they failed to increase GLP-1 levels in the GcgGut-/- mice. Surprisingly, the actions of metformin to increase plasma GLP-1 levels were substantially attenuated in the GcgDistalGut-/- mice. These findings further establish the importance of the proximal gut for the acute response to nutrient-related GLP-1 secretagogues. In contrast, we identify essential contributions of the distal gut to (i) the rapid induction of circulating GLP-1 levels in response to pharmacological selective agonism of G-protein-coupled receptors, (ii) the increased GLP-1 levels following the activation of Toll-Like Receptors with LPS, and iii) the acute GLP-1 response to metformin. Collectively, these results reveal that distal gut Gcg + endocrine cells are rapid responders to structurally and functionally diverse GLP-1 secretagogues.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Dipeptidyl Peptidase IV Inhibitor I
Sigma-Aldrich
Aprotinin from bovine lung, saline solution, 3-7 TIU/mg protein