Merck
  • Colony-stimulating activity from the new metastatic TS/A cell line and its high- and low-metastatic clonal derivatives.
Colony-stimulating activity from the new metastatic TS/A cell line and its high- and low-metastatic clonal derivatives.
British journal of cancer (1985-08-01)
G Nicoletti, P Brambilla, C De Giovanni, P L Lollini, B Del Re, A Marocchi, P Mocarelli, G Prodi, P Nanni
ABSTRACT
We investigated the presence of colony-stimulating factor (CSF) in supernatants obtained from TS/A, a new metastatic murine cell line, and from its high-and low-metastatic clonal derivatives (E and F clones, respectively). TS/A cells produced a CSF in vitro that induced proliferation and differentiation of murine monocytic and granulocytic progenitors in agar cultures. In TS/A-bearing mice remarkable splenomegaly, blood granulocytosis and thymus depletion were observed along with a stimulatory activity in serum and a strong proliferative activity both in spleen and in bone marrow populations. Conditioned media from E clones showed an in vitro colony-stimulating activity greater than those of F clones. Mice injected subcutaneously with cells of all clones studied showed granulocytosis, splenomegaly and thymus depletion, although to varying degrees. However, no direct correlation between granulocytosis-splenomegaly and the number of spontaneous lung metastases was observed.
Sigma-Aldrich
TS/A Mouse Mammary Adenocarcinoma Cell Line, TS/A mouse mammary adenocarcinoma cell line is a well characterized and highly published model for breast cancer research.