Merck
  • Co-expression of B7-1 and ICAM-1 on tumors is required for rejection and the establishment of a memory response.
Co-expression of B7-1 and ICAM-1 on tumors is required for rejection and the establishment of a memory response.
European journal of immunology (1995-05-01)
F Cavallo, A Martin-Fontecha, M Bellone, S Heltai, E Gatti, P Tornaghi, M Freschi, G Forni, P Dellabona, G Casorati
ABSTRACT
Although the transfection of B7-1 cDNA into a few mouse tumor cell lines can induce anti-tumor T cell immunity, its expression alone is ineffective in many other tumor cell lines tested. We were interested to study what factors limit B7-1 co-stimulatory activity, and decided to investigate whether B7-1 requires the cooperation of ICAM-1 to provide the minimal co-stimulatory signal for establishing an efficient anti-tumor immunity. We show that the transfection of B7-1 cDNA into three ICAM-1+ (plasmocytoma J558L, T lymphomas EL-4 and RMA), but not into two ICAM-1- tumors cell lines (adenocarcinoma TS/A and melanoma B16.F1), is sufficient to induce their complete rejection in syngeneic mice. The expression of ICAM-1 is necessary for the rejection of the B7 expressing tumors, since the primary response elicited by B7-1+ EL-4 and RMA clones expressing reduced levels of ICAM-1 is severely reduced. Furthermore, super-transfection of ICAM-1 cDNA into B7-1+ adenocarcinoma and melanoma clones optimizes their primary rejection. Histologic examination of transfected tumors reveals that B7-1 and ICAM-1 exert a potent pro-inflammatory activity. The intra-tumor infiltration is composed of both eosinophils and lymphomonocytes, and is already massive 5 days after the tumor challenge. The primary rejection of the B7-1+ ICAM-1+ tumors depends critically on CD8+ T cells, natural killer cells and granulocytes, but is independent of CD4+ T cells. Remarkably, in addition to its effects on the early phases of the immune response, the co-expression of ICAM-1 and B7-1 on tumors is also necessary for the efficient induction of a memory response. In fact, only the primary challenge with B7-1+, ICAM-1+ tumor cells protects the majority of the mice from a second injection of parental tumor cells. Collectively, our findings indicate that B7-1 and ICAM-1 are fundamental components for triggering the primary rejection of tumors and establishing a protective memory response. These findings may help to define new strategies for the rational application of co-stimulation in tumor immunotherapy.
Sigma-Aldrich
TS/A Mouse Mammary Adenocarcinoma Cell Line, TS/A mouse mammary adenocarcinoma cell line is a well characterized and highly published model for breast cancer research.
Pricing and availability is not currently available. Contact Technical Service