Merck
  • JunB regulates angiogenesis and neurovascular parallel alignment in mouse embryonic skin.
JunB regulates angiogenesis and neurovascular parallel alignment in mouse embryonic skin.
Journal of cell science (2017-01-18)
Yasuo Yoshitomi, Takayuki Ikeda, Hidehito Saito, Yoshino Yoshitake, Yasuhito Ishigaki, Toshihisa Hatta, Nobuo Kato, Hideto Yonekura
ABSTRACT
Blood vessels and nerve fibers are often closely arranged in parallel throughout the body. Therefore, neurovascular interactions have been suggested to be important for the development of vascular networks. However, the molecular mechanisms and genes regulating this process remain unclear. In the present study, we investigated the genes that are activated in endothelial cells (ECs) following interactions with neurons during vascular development. Microarray analyses of human primary microvascular ECs co-cultured with mouse primary dorsal root ganglion cells showed that JunB is strongly upregulated in ECs by neurovascular interactions. Furthermore, the forced expression of JunB in ECs stimulated a tip-like cell formation and angiogenesis in vitro and induced vascular endothelial growth factor A (VEGFA) and the pro-angiogenic integrin subunit ITGB3 expression. Moreover, in vivo knockdown of JunB in ECs from developing mouse limb skin considerably decreased the parallel alignments of blood vessels and nerve fibers. Taken together, the present data demonstrates for the first time that JunB plays an important role in the formation of embryonic vascular networks. These results contribute to the molecular understanding of neurovascular interactions during embryonic vascular development.
Sigma-Aldrich
Anti-Neurofilament 145 kDa Antibody, CT, clone 3H11, clone 3H11, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution