Merck
  • YWHAZ amplification/overexpression defines aggressive bladder cancer and contributes to chemo-/radio-resistance by suppressing caspase-mediated apoptosis.
YWHAZ amplification/overexpression defines aggressive bladder cancer and contributes to chemo-/radio-resistance by suppressing caspase-mediated apoptosis.
The Journal of pathology (2019-04-05)
Chia-Cheng Yu, Chien-Feng Li, I-Hsuan Chen, Ming-Tsung Lai, Zi-Jun Lin, Praveen K Korla, Chee-Yin Chai, Grace Ko, Chih-Mei Chen, Tritium Hwang, Shan-Chih Lee, Jim J-C Sheu
ABSTRACT
The objective of this study was to characterize the oncogenic actions of a recently identified cancer-associated gene YWHAZ (also named as 14-3-3 ζ/δ) in urothelial carcinomas of the urinary bladder (UCUB). A genome-wide study revealed YWHAZ to be involved in the amplicon at 8q22.3, and its genetic amplification was detected predominantly in muscle-invasive bladder cancer (MIBC). Immunohistochemical staining confirmed the association of YWHAZ overexpression with higher tumor stages, lymph node/vascular invasion, and mitotic activity. Univariate and multivariate analyses further indicated the prognostic potential of YWHAZ for more aggressive cancer types. Both gene set enrichment analysis and STRING network studies suggested involvement of YWHAZ in regulating caspase-mediated apoptosis. Ectopic expression of YWHAZ in bladder cells with low endogenous YWHAZ levels boosted cell resistance to doxorubicin and cisplatin, as well as to ionizing radiation. Conversely, YWHAZ-knockdown using specific shRNA in cells with high endogenous YWHAZ levels diminished survival activity, suppressing cell growth and increasing cell death. Our findings confirm the essential role played by YWHAZ in sustaining cell proliferation during chemo/radiotherapy. Treatments based on anti-YWHAZ strategies may thus be beneficial for UCUB patients overexpressing YWHAZ. © 2019 The Authors. The Journal of Pathology published by John Wiley & Sons Ltd on behalf of Pathological Society of Great Britain and Ireland.
Sigma-Aldrich
Anti-PARP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution