Merck
  • MARK4 (Microtubule Affinity-Regulating Kinase 4)-Dependent Inflammasome Activation Promotes Atherosclerosis-Brief Report.
MARK4 (Microtubule Affinity-Regulating Kinase 4)-Dependent Inflammasome Activation Promotes Atherosclerosis-Brief Report.
Arteriosclerosis, thrombosis, and vascular biology (2019-06-07)
Marc Clement, Xiao Chen, Hannah L Chenoweth, Zhongzhao Teng, Sarah Thome, Stephen A Newland, James Harrison, Xian Yu, Alison J Finigan, Ziad Mallat, Xuan Li
ABSTRACT
MARK4 (microtubule affinity-regulating kinase 4) regulates NLRP3 (nucleotide-binding oligomerization domain, leucine-rich repeat, and pyrin domain containing 3) inflammasome activation. The aim of the study is to examine the role of MARK4 in hematopoietic cells during atherosclerosis. We show increased MARK4 expression in human atherosclerotic lesions compared with adjacent areas. MARK4 is coexpressed with NLRP3, and they colocalize in areas enriched in CD68-positive but α-SMA (α-smooth muscle actin)-negative cells. Expression of MARK4 and NLRP3 in the atherosclerotic lesions is associated with the production of active IL (interleukin)-1β and IL-18. To directly assess the role of hematopoietic MARK4 in NLRP3 inflammasome activation and atherosclerotic plaque formation, Ldlr (low-density lipoprotein receptor)-deficient mice were lethally irradiated and reconstituted with either wild-type or Mark4-deficient bone marrow cells, and were subsequently fed a high-fat diet and cholesterol diet for 9 weeks. Mark4 deficiency in bone marrow cells led to a significant reduction of lesion size, together with decreased circulating levels of IL-18 and IFN-γ (interferon-γ). Furthermore, Mark4 deficiency in primary murine bone marrow-derived macrophages prevented cholesterol crystal-induced NLRP3 inflammasome activation, as revealed by reduced caspase-1 activity together with reduced production of IL-1β and IL-18. MARK4-dependent NLRP3 inflammasome activation in the hematopoietic cells regulates the development of atherosclerosis.
Product Description
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Anti-NLRP3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution