
Tetramethyl-Rhodamine-5-dUTP

Synonym(s):
Rhodamine-5-dUTP tetralithium salt, Tetramethylrhodamine-5(6)-(5-[3-carboxamidoallyl]-2′-deoxyuridine 5′-triphosphate)
Empirical Formula (Hill Notation):
C37H40N5O18P3
Molecular Weight:
935.66
Products
Color
Formula Weight
Manufacturer
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing