
Resolve-Al EuFOD

Synonym(s):
Eu(fod)3, Europium(III)-tris(1,1,1,2,2,3,3-heptafluoro-7,7-dimethyl-4,6-octanedionate), Sievers’ Reagent, Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)europium
Linear Formula:
Eu(OCC(CH3)3CHCOCF2CF2CF3)3
Products
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing