
Neosaxitoxin-15N7 solution

Synonym(s):
neo-Saxitoxin-15N7 solution, NeoSTX-15N7
Empirical Formula (Hill Notation):
C10H1715N7O5
Molecular Weight:
322.24
Products
Product Category
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1