
N3-PEG-NH2

Synonym(s):
N3-PEG-NH2
Linear Formula:
N3CH2CH2O(CH2CH2O)nCH2CH2NH2TFA
Products
Brand
Physical Form
Functionality
Shape
Polymer Composition
Polymer Type
Reaction Type
Reactivity
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing