
Maize GMO Standard ERM-BF411

Synonym(s):
GMO Standard ERM-BF411, Maize Bt-176
Products
Manufacturer
Agency
Format
Chemical Composition
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-6 of 6
Compare
Product Number
Description
Pricing