
Dimenhydrinate for peak identification

Synonym(s):
Dimenhydrinate, N-(2-Diphenylmethoxyethyl)-N,N-dimethylammonium 8-chlorotheophyllinate
Empirical Formula (Hill Notation):
C17H21NO · C7H7ClN4O2
CAS Number:
Molecular Weight:
469.96
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing