![Delta9-Tetrahydrocannabinolic acid A solution 1 mg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/253/856/767965bf-c5e1-483e-ad11-c3198bd3ea7a/640/767965bf-c5e1-483e-ad11-c3198bd3ea7a.png)
Delta9-Tetrahydrocannabinolic acid A solution
![Delta9-Tetrahydrocannabinolic acid A solution 1 mg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/253/856/767965bf-c5e1-483e-ad11-c3198bd3ea7a/640/767965bf-c5e1-483e-ad11-c3198bd3ea7a.png)
Synonym(s):
THCA-A
Empirical Formula (Hill Notation):
C22H30O4
CAS Number:
Molecular Weight:
358.47
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing