
Cisatracurium for peak identification

Synonym(s):
Cisatracurium besylate
Empirical Formula (Hill Notation):
C53H72N2O12·2C6H5O3S
CAS Number:
Molecular Weight:
1243.48
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing