
Calcium lactobionate monohydrate

Synonym(s):
Lactobionic acid monohydrate
Empirical Formula (Hill Notation):
C24H42CaO24 · 2H2O
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing