
Bufexamac impurity A

Synonym(s):
4-Butoxyphenylacetic acid
Empirical Formula (Hill Notation):
C12H16O3
CAS Number:
Molecular Weight:
208.25
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing