
Boc-His-OH

Synonym(s):
Nα-(tert-Butoxycarbonyl)-L-histidine, Nα-Boc-L-histidine, Boc-His-OH
Empirical Formula (Hill Notation):
C11H17N3O4
CAS Number:
Molecular Weight:
255.27
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing