
β-Elemonic acid

Synonym(s):
(13α,14β,17α,20S)-3-Oxolanosta-8,24-dien-21-oic acid, 3-Oxotirucalla-8,24-dien-21-oic acid, 3-Oxotirucallic acid, Elemadienonic acid
Empirical Formula (Hill Notation):
C30H46O3
CAS Number:
Molecular Weight:
454.68
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing