
Apamin

Empirical Formula (Hill Notation):
C79H131N31O24S4
CAS Number:
Molecular Weight:
2027.34
Products
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Tissue
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing