
Anti-Bovine Serum antibody produced in rabbit

Synonym(s):
Anti-Serum, bovine
Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Antibody Form
Conjugate
Quality Segment
Package Size
Post Translational Modification
Species Reactivity
Technique