Merck
  • Glycidol, a Valuable Substrate for the Synthesis of Monoalkyl Glyceryl Ethers: A Simplified Life Cycle Approach.
Glycidol, a Valuable Substrate for the Synthesis of Monoalkyl Glyceryl Ethers: A Simplified Life Cycle Approach.
ChemSusChem (2017-04-05)
Maria Ricciardi, Fabrizio Passarini, Ivano Vassura, Antonio Proto, Carmine Capacchione, Raffaele Cucciniello, Daniele Cespi
ABSTRACT
The disposal of any waste by recovering it within the production plant represents the ultimate goal of every biorefinery. In this scenario, the selective preparation of monoalkyl glyceryl ethers (MAGEs) starting from glycidol, obtained as byproduct in the epichlorohydrin production plant, represents a very promising strategy. Here, we report the synthesis of MAGEs through the reaction of glycidol with alcohols catalyzed by a green homogeneous Lewis acids catalyst, such as Bi
Sigma-Aldrich
Bismuth(III) trifluoromethanesulfonate
Pricing and availability is not currently available.
Sigma-Aldrich
Glycidol, 96%
Pricing and availability is not currently available.