Merck
  • Pharmacokinetic-pharmacodynamic study of subcutaneous injection of depot nandrolone decanoate using dried blood spots sampling coupled with ultrapressure liquid chromatography tandem mass spectrometry assays.
Pharmacokinetic-pharmacodynamic study of subcutaneous injection of depot nandrolone decanoate using dried blood spots sampling coupled with ultrapressure liquid chromatography tandem mass spectrometry assays.
The Journal of clinical endocrinology and metabolism (2014-04-02)
Gurmeet K S Singh, Leo Turner, Reena Desai, Mark Jimenez, David J Handelsman
ABSTRACT
Testosterone (T) and nandrolone (N) esters require deep im injections by medical personnel but these often deposit injectate into sc fat so that more convenient sc self-administration may be feasible. To investigate the feasibility and pharmacology of sc injection of N decanoate in healthy men using dried blood spot (DBS) for frequent blood sampling without clinic visits. Healthy male volunteers received 100 mg N decanoate by a single sc injection. Finger-prick capillary blood was spotted onto filter paper before injection daily at home for 21 d and stored at room temperature. Venous whole blood was also spotted onto filter paper before and weekly for 3 wk after injection. DBS were extracted for assay of N and T by liquid chromatography tandem mass spectrometry in a single batch with serum concentrations estimated with adjustment for capillary blood sample volume and hematocrit to define peak (N) or nadir (T) time and concentration from individual daily measurements. Daily serum N peaked 2.50 ± 0.25 (SEM) ng/mL at a median (range) of 6 (4-13) days causing a reduction in serum T from 3.50 ± 0.57 ng/mL at baseline to a nadir of 0.38 ± 0.13 (SEM) ng/mL (89 ± 3% suppression) at a median (range) of 8 (5-16) days. Simultaneously sampled capillary, venous whole blood, and serum gave almost identical results for serum T and N. Finger-pricks and sc injections were well tolerated. This study demonstrates that A) DBS sampling with liquid chromatography mass spectrometry steroid analysis achieves frequent time sampling in the community without requiring clinic visits, venesection, or frozen serum storage, and B) androgen esters in an oil vehicle can be delivered effectively by sc injection, thus avoiding the need for medically supervised deep-im injections.
Product Description
Nandrolone decanoate for peak identification, European Pharmacopoeia (EP) Reference Standard
Nandrolone decanoate for system suitability, European Pharmacopoeia (EP) Reference Standard