Merck
LC-MS-based metabolomics in the clinical laboratory.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2011-11-15)
Susen Becker, Linda Kortz, Christin Helmschrodt, Joachim Thiery, Uta Ceglarek
ABSTRACT
The analysis of metabolites in human body fluids remains a challenge because of their chemical diversity and dynamic concentration range. Liquid chromatography (LC) in combination with tandem mass spectrometry (MS/MS) offers a robust, reliable, and economical methodology for quantitative single metabolite analysis and profiling of complete metabolite classes of a biological specimen over a broad dynamic concentration range. The application of LC-MS/MS based metabolomic approaches in clinical applications aims at both, the improvement of diagnostic sensitivity and specificity by profiling a metabolite class instead of a single metabolite analysis, and the identification of new disease specific biomarkers. In the present paper we discuss recent advances in method development for LC-MS/MS analysis of lipids, carbohydrates, amino acids and biogenic amines, vitamins and organic acids with focus on human body fluids. In this context an overview on recent LC-MS/MS based metabolome studies for cancer, diabetes and coronary heart disease is presented.
Sigma-Aldrich
D-Xylulose 5-phosphate sodium salt, ≥95% (TLC)
Sigma-Aldrich
D-Xylulose 5-phosphate lithium salt, ≥90% (TLC)