Merck
Effects of desogestrel on carbohydrate metabolism.
American journal of obstetrics and gynecology (1993-03-01)
D Shoupe
ABSTRACT
Associated with progesterone and the synthetic progestins used in oral contraceptives is a dose-dependent impairment of carbohydrate metabolism. It is well known that in the general population hyperinsulinemia and alterations in glucose metabolism are significant risk factors for the development of cardiovascular disease. Studies that use curve analysis of glucose tolerance tests have demonstrated insulin resistance, rises in plasma insulin, and relative glucose intolerance in women using oral contraceptives. Desogestrel, a new progestin, has been demonstrated to have generally less pronounced effects on these parameters of carbohydrate metabolism.
Product Description
Desogestrel for system suitability, European Pharmacopoeia (EP) Reference Standard
Desogestrel, European Pharmacopoeia (EP) Reference Standard