Merck
  • Palladium-catalyzed sp2 and sp3 C-H bond activation and addition to isatin toward 3-hydroxy-2-oxindoles.
Palladium-catalyzed sp2 and sp3 C-H bond activation and addition to isatin toward 3-hydroxy-2-oxindoles.
Organic letters (2013-10-05)
Gang-Wei Wang, An-Xi Zhou, Jun-Jiao Wang, Rong-Bin Hu, Shang-Dong Yang
ABSTRACT
The first Pd(II)-catalyzed C-H addition to isatins by direct sp(2)/sp(3) C-H bond activation for the construction of 3-substituted-3-hydroxy-2-oxindoles is reported. The bidentate nitrogen ligands were found to promote this reaction. Specifically, the preliminary bioassay indicated that 3-(5-chlorobenzoxazole)-3-hydroxy-N-benzyl-2-oxindole (2w) is a new inhibitor of human kidney cancer and hepatocellular carcinoma cells. Moreover, this reaction system exhibits great functional group tolerance and requires no directing group, extra base, or additives.
Sigma-Aldrich
Isatin, 97%
Pricing and availability is not currently available.