Merck
  • Halogen-metal exchange in 1,2-dibromobenzene and the possible intermediacy of 1,2-dilithiobenzene.
Halogen-metal exchange in 1,2-dibromobenzene and the possible intermediacy of 1,2-dilithiobenzene.
The Journal of organic chemistry (2007-11-08)
Holger F Bettinger, Matthias Filthaus
ABSTRACT
The one-step high-yield synthesis of 1,2-bis(trimethylsilyl)benzene from 1,2-dibromobenzene using tert-butyllithium and trimethylsilyltriflate is reported. A mechanistic investigation shows that 1,2-dilithiobenzene is not an intermediate in this reaction; the coexistence of trimethylsilyltriflate and tert-butyllithium at very low temperatures allows a sequence of bromine-lithium exchange and subsequent derivatization reactions to operate.
Sigma-Aldrich
1,2-Dibromobenzene, 98%
Pricing and availability is not currently available.