Merck
  • Induction of aneuploidy in Chinese hamster oocytes following in vivo treatments with trimethoxybenzoic compounds and their analogues.
Induction of aneuploidy in Chinese hamster oocytes following in vivo treatments with trimethoxybenzoic compounds and their analogues.
Mutation research (1995-03-01)
H Tateno, Y Kamiguchi, M Shimada, S Sugawara, K Mikamo
ABSTRACT
Many inhibitors of tubulin polymerization have a trimethoxybenzene ring in their molecules. Such trimethoxybenzoic compounds and their analogues may therefore have a potency to induce meiotic nondisjunction of oocytes. In this study, a single dose of reserpine (0.5 microgram/g body weight), podophyllotoxin (20.0 micrograms/g b.w.), trimethoxybenzoic acid (500.0 micrograms/g b.w.) or vinblastine sulfate (3.0 micrograms/g b.w.) was injected intraperitoneally to mature female Chinese hamsters at the onset of the first meiotic spindle formation of oocytes. Within 6 h after spontaneous ovulation, MII oocytes were collected from the oviducts for morphological examination and cytogenetic analysis. The incidence of morphologically abnormal oocytes with unusually large first polar body or bodies increased significantly after the treatment with reserpine (18/202; 8.9%), podophyllotoxin (28/172; 16.3%) and vinblastine sulfate (63/197; 32.0%), as compared with the control (3/214; 1.4%). Chromosome analysis of oocytes revealed that podophyllotoxin and vinblastine sulfate were effective in inducing aneuploidy (62/154; 40.3% and 128/156; 82.1% vs. 3/198; 1.5% of the control) by inhibiting the formation of spindle microtubules at the first meiosis. Aneuploids were found more frequently in morphologically abnormal oocytes than in normal oocytes. No aneugenic activity of reserpine and trimethoxybenzoic acid was observed. These results indicate that trimethoxybenzoic compounds do not necessarily exhibit aneugenic activity.
Sigma-Aldrich
3,4,5-Trimethoxybenzoic acid, ReagentPlus®, 99%
Pricing and availability is not currently available.