Merck
  • Chrysin: a histone deacetylase 8 inhibitor with anticancer activity and a suitable candidate for the standardization of Chinese propolis.
Chrysin: a histone deacetylase 8 inhibitor with anticancer activity and a suitable candidate for the standardization of Chinese propolis.
Journal of agricultural and food chemistry (2012-11-09)
Li-Ping Sun, Ai-Ling Chen, Hsiao-Chiao Hung, Yin-Huan Chien, Jing-Shi Huang, Chung-Yang Huang, Yue-Wen Chen, Chia-Nan Chen
ABSTRACT
Chinese propolis (CP) is a natural product collected by honeybees and a health food raw material. Previous studies have shown that CP exhibits a broad spectrum of biological activities including anticancer, antioxidant, antibacterial, anti-inflammatory, and antiviral activities. The focuses of the present study were the standardization of CP and the possible mechanisms of its active anticancer ingredients. Nine samples of CP were collected from different locations in China. Analyses of the CP samples revealed that all 9 had similar chemical compositions. Parameters analyzed included the CP extract dry weight, total phenolic content, and DPPH free radical scavenging activities. The active anticancer ingredient was isolated, characterized against human MDA-MB-231 breast cancer cells, and identified as chyrsin, a known potent anticancer compound. Chrysin is present at high levels in all 9 of the CP samples, constituting approximately 2.52% to 6.38% of the CP extracts. However, caffeic acid phenethyl ester (CAPE), another potent active ingredient is present in low levels in 9 samples of CP, constituting approximately 0.08% to 1.71% of the CP extracts. Results from analyses of enzymatic activity indicated that chrysin is a histone deacetylase inhibitor (HDACi) and that it markedly inhibited HDAC8 enzymatic activity (EC(50) = 40.2 μM). In vitro analyses demonstrated that chrysin significantly suppressed cell growth and induced differentiation in MDA-MB-231 cells. In a xenograft animal model (MDA-MB-231 cells), orally administered chrysin (90 mg/kg/day) significantly inhibited tumor growth. Despite the geographical diversity of the 9 samples' botanical origins, their chemical compositions and several analyzed parameters were similar, suggesting that CP is standardized, with chrysin being the major active ingredient. Overall, in vitro and in vivo data indicated that chrysin is an HDAC8 inhibitor, which can significantly inhibit tumor growth. Data also suggested that chrysin might represent a suitable candidate for standardization of CP.
Product Description
Supelco
Chrysin, analytical standard
Sigma-Aldrich
Chrysin, ≥96.5%