Merck
  • Disila-analogues of the synthetic retinoids EC23 and TTNN: synthesis, structure and biological evaluation.
Disila-analogues of the synthetic retinoids EC23 and TTNN: synthesis, structure and biological evaluation.
Organic & biomolecular chemistry (2012-07-21)
Josef B G Gluyas, Christian Burschka, Steffen Dörrich, Judith Vallet, Hinrich Gronemeyer, Reinhold Tacke
ABSTRACT
Silicon chemistry offers the potential to tune the effects of biologically active organic molecules. Subtle changes in the molecular backbone caused by the exchange of a carbon atom for a silicon atom (sila-substitution) can significantly alter the biological properties. In this study, the biological effects of a two-fold sila-substitution in the synthetic retinoids EC23 (4-(5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-ylethynyl)benzoic acid (4a)) and TTNN (6-(5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)-2-naphthoic acid (7a)) as well as their corresponding analogues with an indane instead of a 1,2,3,4-tetrahydronaphthalene skeleton (compounds 5a and 8a) were investigated. Two-fold C/Si exchange in 4a, 5a, 7a and 8a leads to the silicon-analogues disila-EC23 (4b), 5b, disila-TTNN (7b) and 8b, which contain a 1,2,3,4-tetrahydro-1,4-disilanaphthalene (4b, 7b) or 1,3-disilaindane skeleton (5b, 8b). Exchange of the SiCH(2)Si moiety of 5b for an SiOSi fragment leads to the disiloxane 6 (2-oxa-1,3-disilaindane skeleton). The EC23 derivative 5a, the TTNN derivative 8a and the silicon-containing analogues 4b, 5b, 6, 7b and 8b were synthesised, and the biological properties of the C/Si pairs 4a/4b, 5a/5b, 7a/7b and 8a/8b and compound 6 were evaluated in vivo using RAR isotype-selective reporter cells. EC23 (4a) and its derivatives disila-EC23 (4b), 5a, 5b and 6 are very potent RAR agonists, which are even more potent than the powerful reference compound TTNPB. Disila-substitution of EC23 (4a) and 5a leads to a moderate decrease in RARα activation, whereas the RARβ,γ activation is almost not affected. In contrast, two-fold C/Si exchange in the weak retinoid agonist TTNN (7a) and 8a resulted in considerably different effects: a significant increase (7a→7b) and almost no change (8a→8b) in transcription activation potential for all three RAR isotypes. Disila-TTNN (7b) can be regarded as a powerful RARβ,γ-selective retinoid.
Sigma-Aldrich
1-Naphthoic acid, 96%
Pricing and availability is not currently available.