Merck
  • A route to 1,4-disubstituted aromatics and its application to the synthesis of the antibiotic culpin.
A route to 1,4-disubstituted aromatics and its application to the synthesis of the antibiotic culpin.
The Journal of organic chemistry (2008-09-26)
Rajesh Sunasee, Derrick L J Clive
ABSTRACT
A method is described for converting tert-butyl benzoates or tert-butyl 1-naphthoates into derivatives having an alkyl or substituted alkyl group in a 1,4-relationship to an alkyl, aryl, alkenyl, or alkynyl group. Key steps in the sequence are (i) addition of an organometallic species to a cross-conjugated cyclohexadienone obtained by Birch alkylation of a tert-butyl benzoate or a tert-butyl 1-naphthoate, followed by allylic oxidation, and (ii) treatment with BiCl3 x H2O, which results in removal of the tert-butyl group and spontaneous decarboxylative aromatization. The method was applied to the synthesis of the antimicrobial fungal metabolite culpin.
Sigma-Aldrich
1-Naphthoic acid, 96%
Pricing and availability is not currently available.