- [Gynalgin--one more opportunity in the treatment of bacterial vaginosis].
[Gynalgin--one more opportunity in the treatment of bacterial vaginosis].
The data of 30 nonpregnant women in reproductive age BV treated with Gynalgin were presented. On clinical and microbiological indicator Gynalgin showed a good effect on BV-associated microorganisms (80%). Only one of the cases of BV was not affected (4%). With nixed infection of BV and Candida we established clinical improvement and recovery of Lactobacillus spp. In 66.6%, but in all cases on Candida is not effect. We consider that Gynalgin is still one more easily applicable and clinically effective opportunity for treatment of BV.