Merck
  • Commercially derived versatile optical architecture for two-photon STED, wavelength mixing and label-free microscopy.
Commercially derived versatile optical architecture for two-photon STED, wavelength mixing and label-free microscopy.
Biomedical optics express (2022-04-14)
Chiara Peres, Chiara Nardin, Guang Yang, Fabio Mammano
ABSTRACT
Multimodal microscopy combines multiple non-linear techniques that take advantage of different optical processes to generate contrast and increase the amount of information that can be obtained from biological samples. However, the most advanced optical architectures are typically custom-made and often require on-site adjustment of optical components performed by trained personnel for optimal performance. Here, we describe a hybrid system we built based on a commercial upright microscope. We show that our multimodal imaging platform can be used to seamlessly perform two-photon STED, wavelength mixing and label-free microscopy in both ex vivo and in vivo turbid samples. The system is stable and endowed with remote alignment hardware that ensures long-term operability also for non-expert users, using the alignment protocol described in this article and in the related material. This optical architecture is an important step forward towards a wider practical applicability of non-linear optics to bioimaging.
Sigma-Aldrich
Anti-Tom20/Tomm20 Antibody, clone 2F8.1, clone 2F8.1, from mouse
Pricing and availability is not currently available.