Merck
  • Synthesis of Highly Conductive Electrospun Recycled Polyethylene Terephthalate Nanofibers Using the Electroless Deposition Method.
Synthesis of Highly Conductive Electrospun Recycled Polyethylene Terephthalate Nanofibers Using the Electroless Deposition Method.
Nanomaterials (Basel, Switzerland) (2021-03-07)
Nadir Hussain, Mujahid Mehdi, Muhammad Yousif, Aizaz Ali, Sana Ullah, Sajid Hussain Siyal, Tanweer Hussain, Ick Soo Kim
ABSTRACT
Plastic bottles are generally recycled by remolding them into numerous products. In this study, waste from plastic bottles was used to fabricate recycled polyethylene terephthalate (r-PET) nanofibers via the electrospinning technique, and high-performance conductive polyethylene terephthalate nanofibers (r-PET nanofibers) were prepared followed by copper deposition using the electroless deposition (ELD) method. Firstly, the electrospun r-PET nanofibers were chemically modified with silane molecules and polymerized with 2-(methacryloyloxy) ethyl trimethylammonium chloride (METAC) solution. Finally, the copper deposition was achieved on the surface of chemically modified r-PET nanofibers by simple chemical/ion attraction. The water contact angle of r-PET nanofibers, chemically modified r-PET nanofibers, and copper deposited nanofibers were 140°, 80°, and 138°, respectively. The r-PET nanofibers retained their fibrous morphology after copper deposition, and EDX results confirmed the presence of copper on the surface of r-PET nanofibers. XPS was performed to analyze chemical changes before and after copper deposition on r-PET nanofibers. The successful deposition of copper one r-PET nanofibers showed an excellent electrical resistance of 0.1 ohms/cm and good mechanical strength according to ASTM D-638.
Sigma-Aldrich
Vinyltrimethoxysilane, 98%
Pricing and availability is not currently available.