Merck
  • Omics-Driven Systems Interrogation of Metabolic Dysregulation in COVID-19 Pathogenesis.
Omics-Driven Systems Interrogation of Metabolic Dysregulation in COVID-19 Pathogenesis.
Cell metabolism (2020-07-02)
Jin-Wen Song, Sin Man Lam, Xing Fan, Wen-Jing Cao, Si-Yu Wang, He Tian, Gek Huey Chua, Chao Zhang, Fan-Ping Meng, Zhe Xu, Jun-Liang Fu, Lei Huang, Peng Xia, Tao Yang, Shaohua Zhang, Bowen Li, Tian-Jun Jiang, Raoxu Wang, Zehua Wang, Ming Shi, Ji-Yuan Zhang, Fu-Sheng Wang, Guanghou Shui
ABSTRACT
The coronavirus disease 2019 (COVID-19) pandemic presents an unprecedented threat to global public health. Herein, we utilized a combination of targeted and untargeted tandem mass spectrometry to analyze the plasma lipidome and metabolome in mild, moderate, and severe COVID-19 patients and healthy controls. A panel of 10 plasma metabolites effectively distinguished COVID-19 patients from healthy controls (AUC = 0.975). Plasma lipidome of COVID-19 resembled that of monosialodihexosyl ganglioside (GM3)-enriched exosomes, with enhanced levels of sphingomyelins (SMs) and GM3s, and reduced diacylglycerols (DAGs). Systems evaluation of metabolic dysregulation in COVID-19 was performed using multiscale embedded differential correlation network analyses. Using exosomes isolated from the same cohort, we demonstrated that exosomes of COVID-19 patients with elevating disease severity were increasingly enriched in GM3s. Our work suggests that GM3-enriched exosomes may partake in pathological processes related to COVID-19 pathogenesis and presents the largest repository on the plasma lipidome and metabolome distinct to COVID-19.
Sigma-Aldrich
Sodium L-lactate-13C3 solution, 45-55 % (w/w) in H2O, ≥99 atom % 13C, ≥98% (CP), ≥98% (Chiral purity, HPLC)