Merck
  • Arrhythmogenesis toxicity of aconitine is related to intracellular ca(2+) signals.
Arrhythmogenesis toxicity of aconitine is related to intracellular ca(2+) signals.
International journal of medical sciences (2013-08-13)
Yu-hong Zhou, Xian-mei Piao, Xue Liu, Hai-hai Liang, Lei-min Wang, Xue-hui Xiong, Lu Wang, Yan-jie Lu, Hong-li Shan
ABSTRACT
Aconitine is a well-known arrhythmogenic toxin and induces triggered activities through cardiac voltage-gated Na(+) channels. However, the effects of aconitine on intracellular Ca(2+) signals were previously unknown. We investigated the effects of aconitine on intracellular Ca(2+) signals in rat ventricular myocytes and explored the possible mechanism of arrhythmogenic toxicity induced by aconitine. Ca(2+) signals were evaluated by measuring L-type Ca(2+) currents, caffeine-induced Ca(2+) release and the expression of NCX and SERCA2a. Action potential and triggered activities were recorded by whole-cell patch-clamp techniques. In rat ventricular myocytes, the action potential duration was significantly prolonged by 1 µM aconitine. At higher concentrations (5 µM and 10 µM), aconitine induced triggered activities and delayed after-depolarizations (6 of 8 cases), which were inhibited by verapamil. Aconitine (1 µM) significantly increased the ICa-L density from 12.77 ± 3.12 pA/pF to 18.98 ± 3.89 pA/pF (n=10, p<0.01). The activation curve was shifted towards more negative potential, while the inactivation curve was shifted towards more positive potential by 1 μM aconitine. The level of Ca(2+) release induced by 10 mM caffeine was markedly increased. Aconitine (1 µM) increased the expression of NCX, while SERCA2a expression was reduced. In conclusion, aconitine increased the cytosolic [Ca(2+)]i by accelerating ICa-L and changing the expression of NCX and SERCA2a. Then, the elevation of cytosolic [Ca(2+)]i induced triggered activities and delayed after-depolarizations. Arrhythmogenesis toxicity of aconitine is related to intracellular Ca(2+) signals.
Sigma-Aldrich
Aconitine, ≥95% (HPLC), crystalline