Merck
  • Differential Anti-Proliferative and Anti-Migratory Activities of Ursolic Acid, 3-O-Acetylursolic Acid and Their Combination Treatments with Quercetin on Melanoma Cells.
Differential Anti-Proliferative and Anti-Migratory Activities of Ursolic Acid, 3-O-Acetylursolic Acid and Their Combination Treatments with Quercetin on Melanoma Cells.
Biomolecules (2020-06-18)
Aljawharah AlQathama, Luying Shao, Ammar Bader, Proma Khondkar, Simon Gibbons, Jose M Prieto
ABSTRACT
We evaluate how 3-acetylation modulates the in vitro activity of ursolic acid in melanoma cells alone or in combination treatments with quercetin. Anti-proliferative studies on A375 cells and adult human dermal fibroblasts included analyses on cell cycle distribution, caspase activity, phosphatidylserine translocation, cell morphology and Bax/Bcl-2 protein expression. Then, 2D and 3D migration of B16F10 cells were studied using scratch and Transwell assays, respectively. Ursolic acid and 3-O-acetylursolic acid have shown similar GI50 on A375 cells (26 µM vs. 32 µM, respectively) significantly increased both early and late apoptotic populations, activated caspases 3/7 (48-72 h), and enhanced Bax whilst attenuating Bcl-2 expression. Ursolic acid caused elevation of the sub-G1 population whilst its 3-acetyl derivative arrested cell cycle at S phase and induced strong morphological changes. Combination treatments showed that ursolic acid and quercetin act synergistically in migration assays but not against cell proliferation. In summary, 3-O-acetylursolic acid maintains the potency and overall apoptotic mechanism of the parent molecule with a more aggressive influence on the morphology of A375 melanoma cells but the 3-acetylation suppresses its anti-migratory properties. We also found that ursolic acid can act in synergy with quercetin to reduce cell migration.
Sigma-Aldrich
Ursolic acid, ≥90%
Pricing and availability is not currently available.