Merck
  • Visible-Light-Mediated Iodoperfluoroalkylation of Alkenes in Flow and Its Application to the Synthesis of a Key Fulvestrant Intermediate.
Visible-Light-Mediated Iodoperfluoroalkylation of Alkenes in Flow and Its Application to the Synthesis of a Key Fulvestrant Intermediate.
Organic letters (2019-06-30)
Cristian Rosso, Jason D Williams, Giacomo Filippini, Maurizio Prato, C Oliver Kappe
ABSTRACT
Two efficient continuous flow iodoperfluoroalkylation methods are described: using 0.05 mol % perylene diimide (PDI) photocatalyst under 450 nm irradiation or substoichiometric triethylamine under 405 nm irradiation. These methods enable dramatically elevated productivity versus batch processes. The triethylamine-mediated method is explored mechanistically and in substrate scope. The gram-scale synthesis of an active pharmaceutical ingredient side chain is also reported in flow, via a photochemical iodoperfluoroalkylation followed by hydrogenolysis.
Sigma-Aldrich
iPr-PDI, ≥95%
Pricing and availability is not currently available.