Merck
  • The impact of technical failures during cultivation of an inclusion body process.
The impact of technical failures during cultivation of an inclusion body process.
Bioprocess and biosystems engineering (2019-07-04)
Alexander Pekarsky, Vanessa Konopek, Oliver Spadiut
ABSTRACT
In biotechnological processes, technical failures in the upstream process often lead to batch loss. It is of great interest to investigate the empirical impact of technical failures to understand and mitigate their impact accurately and reduce economic damage. We investigated the impact in the upstream and downstream of a recombinant antibody fragment inclusion body production process chain to provide integrated empirical data and knowledge. First, we provided a reproducible process chain that yielded high inclusion body content, high specific product titer, and a refolding yield of 30%. The inclusion body downstream proved to be of high reproducibility. Through the intended introduction of technical failures, we were not only able to shed more light on the empirical responses in the upstream and downstream, but also on process-boosting parameters that would have been neglected. Herein, a short increase in temperature during the cultivation clearly increased the refolding yield.
Sigma-Aldrich
Protein standard, Micro Standard, liquid
Pricing and availability is not currently available.