Merck
  • Playing the DISC: turning on TRAIL death receptor-mediated apoptosis in cancer.
Playing the DISC: turning on TRAIL death receptor-mediated apoptosis in cancer.
Biochimica et biophysica acta (2009-12-08)
Bodvaël Pennarun, Annemieke Meijer, Elisabeth G E de Vries, Jan H Kleibeuker, Frank Kruyt, Steven de Jong
ABSTRACT
Formation of the pro-apoptotic death-inducing signaling complex (DISC) can be initiated in cancer cells via binding of tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) to its two pro-apoptotic receptors, TRAIL receptor 1 (TRAIL-R1) and TRAIL-R2. Primary components of the DISC are trimerized TRAIL-R1/-R2, FADD, caspase 8 and caspase 10. The anti-apoptotic protein FLIP can also be recruited to the DISC to replace caspase 8 and form an inactive complex. Caspase 8/10 processing at the DISC triggers the caspase cascade, which eventually leads to apoptotic cell death. Besides TRAIL, TRAIL-R1- or TRAIL-R2-selective variants of TRAIL and agonistic antibodies have been designed. These ligands are of interest as anti-cancer agents since they selectively kill tumor cells. To increase tumor sensitivity to TRAIL death receptor-mediated apoptosis and to overcome drug resistance, TRAIL receptor ligands have already been combined with various therapies in preclinical models. In this review, we discuss factors influencing the initial steps of the TRAIL apoptosis signaling pathway, focusing on mechanisms modulating DISC assembly and caspase activation at the DISC. These insights will direct rational design of drug combinations with TRAIL receptor ligands to maximize DISC signaling.
Sigma-Aldrich
Sodium glycochenodeoxycholate, ≥97.0% (TLC)
Pricing and availability is not currently available.