Silver(I) chloride
Synonym(s):
Silver chloride
Empirical Formula (Hill Notation):
AgCl
CAS Number:
Molecular Weight:
143.32
Products
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing