Skip to Content
Merck
Potassium hexachloroplatinate(IV) ≥99.99% trace metals basis

Potassium hexachloroplatinate(IV)

Potassium hexachloroplatinate(IV) ≥99.99% trace metals basis
Synonym(s):
Dipotassium hexachloroplatinate, Dipotassium platinum hexachloride, Platinic potassium chloride, Platinum potassium chloride, Potassium chloroplatinate, Potassium platinum(IV) chloride, Potassium platinum(IV) hexachloride
Linear Formula:
K2PtCl6
CAS Number:
Molecular Weight:
485.99
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Chemical Composition
Reagent Type
Core