
Perindopril for peak identification

Synonym(s):
Perindopril erbumine, (2S,3aS,7aS)-1-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)butyl]amino]-1-oxopropyl]octahydro-1H-indole-2-carboxylic acid tert-butylamine salt, Perindopril tert-butylamine, S-9490
Empirical Formula (Hill Notation):
C19H32N2O5 · C4H11N
CAS Number:
Molecular Weight:
441.60
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing