
Octreotide

Synonym(s):
Octreotide acetate salt
Empirical Formula (Hill Notation):
C49H66N10O10S2 · xC2H4O2
CAS Number:
Molecular Weight:
1019.24 (free base basis)
Products
Formula Weight
Manufacturer
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing