
Hydrochlorothiazide for peak identification

Synonym(s):
Hydrochlorothiazide, 6-Chloro-3,4-dihydro-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide, 6-Chloro-7-sulfamyl-3,4-dihydro-1,2,4-benzothiadiazine 1,1-dioxide
Empirical Formula (Hill Notation):
C7H8ClN3O4S2
CAS Number:
Molecular Weight:
297.74
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing