
(±)-Jasmonic acid

Empirical Formula (Hill Notation):
C12H18O3
CAS Number:
Molecular Weight:
210.27
Products
Brand
Formula Weight
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing