Skip to Content
Merck
Decanoic acid analytical standard

Decanoic acid

Decanoic acid analytical standard
Synonym(s):
1-Nonanecarboxylic acid, Acid C10, C10:0, Capric acid, NSC 5025
Linear Formula:
CH3(CH2)8COOH
CAS Number:
Molecular Weight:
172.26
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing