
Cynarin

Synonym(s):
1,3-Dicaffeoylquinic acid, (1R,3R,4S,5R)-1,3-Bis[[3-(3,4-dihydroxyphenyl)-1-oxo-2-propen-1-yl]oxy]-4,5-dihydroxycyclohexanecarboxylic acid, 1,5-DQA, 1,5-Dicaffeoylquinic acid, Cynarin, Cynarine
Empirical Formula (Hill Notation):
C25H24O12
CAS Number:
Molecular Weight:
516.45
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing